Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015976.4 | NP_057060.2 | 4 | 630 | ||
NR_033716.1 | 5 | 750 | ||||
XM_006710677.2 | XP_006710740.1 | 4 | 647 | |||
XM_006710678.2 | XP_006710741.1 | 5 | 746 | |||
XM_011541564.2 | XP_011539866.1 | 5 | 686 | |||
XM_017001425.1 | XP_016856914.1 | 4 | 608 | |||
XM_017001426.1 | XP_016856915.1 | 5 | 745 | |||
XM_017001427.1 | XP_016856916.1 | 5 | 1020 | |||
XM_017001428.1 | XP_016856917.1 | 4 | 495 | |||
GenBank mRNA | AF121857.1 | - | 4 | 473 | ||
AK025031.1 | - | 4 | 587 | |||
AK223229.1 | - | 4 | 489 | |||
AK293018.1 | - | 4 | 558 | |||
AL049989.1 | - | 4 | 487 | |||
AL548028.3 | - | 4 | 631 | |||
BC010349.1 | - | 4 | 444 | 4093672 | ||
BC018105.1 | - | 5 | 568 | 3849824 | ||
BT006747.1 | - | 3 | 386 |