Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244705.1 | NP_001231634.1 | 16 | 1554 | ||
NM_001244706.1 | NP_001231635.1 | 7 | 696 | |||
NM_015989.4 | NP_057073.4 | 16 | 1651 | |||
XM_011538442.2 | XP_011536744.2 | 16 | 2203 | |||
XM_011538446.2 | XP_011536748.1 | 15 | 2405 | |||
XM_011538449.1 | XP_011536751.1 | 10 | 773 | |||
XM_011538451.1 | XP_011536753.1 | 12 | 1074 | |||
XM_017019416.1 | XP_016874905.1 | 13 | 1796 | |||
XM_017019417.1 | XP_016874906.1 | 13 | 1171 | |||
XM_017019418.1 | XP_016874907.1 | 12 | 1184 | |||
XM_017019419.1 | XP_016874908.1 | 11 | 1028 | |||
GenBank mRNA | AB044561.1 | - | 7 | 696 | ||
AF116545.1 | - | 15 | 1394 | |||
AF116546.1 | - | 11 | 949 | |||
AF116547.1 | - | 15 | 1362 | |||
AF116548.1 | - | 16 | 1601 | |||
AK122681.1 | - | 14 | 2108 | |||
AK289659.1 | - | 11 | 1070 | |||
AK315265.1 | - | 16 | 1578 | |||
BC052249.1 | - | 16 | 1554 | |||
BC098278.1 | - | 15 | 1427 | 40006057 | ||
BC098342.1 | - | 15 | 1427 | 40006051 | ||
BC099717.1 | - | 15 | 1427 | 40006058 | ||
BC105918.1 | - | 15 | 1427 | 40006053 |