Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001220488.1 | NP_001207417.1 | 13 | 2240 | ||
NM_001220489.1 | NP_001207418.1 | 11 | 2017 | |||
NM_001220490.1 | NP_001207419.1 | 11 | 1925 | |||
NM_001257.4 | NP_001248.1 | 12 | 2134 | |||
XM_011522804.2 | XP_011521106.1 | 11 | 1741 | |||
GenBank mRNA | AK294277.1 | - | 6 | 997 | ||
AK295886.1 | - | 11 | 1755 | |||
AK298612.1 | - | 11 | 1843 | |||
AK304592.1 | - | 5 | 633 | |||
AK304733.1 | - | 13 | 1995 | |||
BC028624.2 | - | 12 | 1926 | 4825734 | ||
BC030653.2 | - | 12 | 1926 | 4811561 | ||
BG911233.1 | - | 1 | 22 | |||
L34058.1 | - | 12 | 2279 | |||
U59288.1 | - | 12 | 2051 |