Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016101.4 | NP_057185.1 | 4 | 681 | ||
GenBank mRNA | AB062398.1 | - | 4 | 383 | ||
AB062479.1 | - | 4 | 383 | |||
AB112439.2 | - | 4 | 521 | |||
AF085360.1 | - | 4 | 396 | |||
AF132971.1 | - | 4 | 396 | |||
AK315355.1 | - | 4 | 414 | |||
BC015941.1 | - | 4 | 408 | 3922688 |