Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016113.4 | NP_057197.2 | 5 | 1159 | ||
XM_005256676.1 | XP_005256733.1 | 5 | 1159 | |||
XM_005256677.1 | XP_005256734.1 | 5 | 1159 | |||
XM_005256678.4 | XP_005256735.1 | 5 | 1159 | |||
XM_006721541.3 | XP_006721604.1 | 5 | 1159 | |||
XM_006721543.3 | XP_006721606.1 | 5 | 1159 | |||
XM_011523922.1 | XP_011522224.1 | 5 | 1159 | |||
XM_011523923.1 | XP_011522225.1 | 5 | 1159 | |||
XM_017024730.1 | XP_016880219.1 | 5 | 1159 | |||
XM_017024731.1 | XP_016880220.1 | 5 | 1159 | |||
XM_017024732.1 | XP_016880221.1 | 5 | 1159 | |||
GenBank mRNA | AF103906.1 | - | 5 | 925 | ||
AF129112.1 | - | 4 | 795 | |||
AJ487963.1 | - | 4 | 764 | |||
AK289705.1 | - | 5 | 1159 | |||
BC018926.2 | - | 5 | 1127 | 4298484 | ||
BC051305.1 | - | 5 | 1115 | 4870010 |