Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308080.1 | NP_001295009.1 | 2 | 214 | ||
NM_016144.3 | NP_057228.1 | 2 | 187 | |||
XM_011543458.2 | XP_011541760.1 | 2 | 571 | |||
XM_011543460.2 | XP_011541762.1 | 2 | 570 | |||
XM_017009553.1 | XP_016865042.1 | 2 | 309 | |||
XM_017009555.1 | XP_016865044.1 | 2 | 569 | |||
XM_017009556.1 | XP_016865045.1 | 2 | 569 | |||
GenBank mRNA | AF078857.1 | - | 2 | 142 | ||
AF161423.1 | - | 2 | 138 | |||
AK002147.1 | - | 2 | 163 | |||
AY542165.1 | - | 2 | 110 | |||
BC005179.1 | - | 2 | 133 | 3683093 | ||
BI859871.1 | - | 2 | 203 | |||
DA001572.1 | - | 2 | 214 | |||
HY139221.1 | - | 2 | 188 |