Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR83OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016145.3 | NP_057229.1 | 4 | 828 | ||
XM_017026864.1 | XP_016882353.1 | 5 | 1822 | |||
GenBank mRNA | AF059620.1 | - | 4 | 596 | ||
AF078861.1 | - | 4 | 813 | |||
AF151898.1 | - | 4 | 812 | |||
AK091030.1 | - | 4 | 2533 | |||
BC001192.1 | - | 4 | 576 | |||
BC042919.1 | - | 4 | 597 | 5932109 | ||
BC061912.1 | - | 4 | 597 | 4444565 |