Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMOTL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278683.1 | NP_001265612.1 | 4 | 1464 | ||
NM_001278685.1 | NP_001265614.1 | 4 | 1478 | |||
NM_016201.3 | NP_057285.3 | 4 | 1478 | |||
XM_006713654.2 | XP_006713717.1 | 4 | 1496 | |||
XM_011512881.1 | XP_011511183.1 | 4 | 1335 | |||
XM_017006580.1 | XP_016862069.1 | 4 | 1335 | |||
XM_017006581.1 | XP_016862070.1 | 4 | 1516 | |||
XM_017006582.1 | XP_016862071.1 | 4 | 1343 | |||
GenBank mRNA | AB023206.1 | - | 4 | 1411 | ||
AF175966.1 | - | 3 | 545 | |||
AK001343.1 | - | 4 | 1341 | |||
AK091040.1 | - | 1 | 546 | |||
AK223598.1 | - | 4 | 1336 | |||
AK291616.1 | - | 4 | 1422 | |||
AK299270.1 | - | 4 | 1464 | |||
BC011454.1 | - | 2 | 444 | 4181482 | ||
BC025981.2 | - | 1 | 34 |