Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUCO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282751.1 | NP_001269680.1 | 16 | 2914 | ||
NM_014283.4 | NP_055098.1 | 18 | 3074 | |||
NM_016227.3 | NP_057311.3 | 17 | 3333 | |||
XM_006711374.2 | XP_006711437.1 | 17 | 3134 | |||
XM_006711375.2 | XP_006711438.1 | 17 | 3044 | |||
XM_006711376.1 | XP_006711439.1 | 16 | 3023 | |||
GenBank mRNA | AF097535.1 | - | 17 | 3333 | ||
AJ250075.1 | - | 18 | 3072 | |||
AK302949.1 | - | 18 | 3074 | |||
AL035291.1 | - | 18 | 2975 | |||
BC137125.1 | - | 18 | 2860 | 9021122 | ||
BC144487.1 | - | 16 | 2700 |