Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137559.1 | NP_001131031.1 | 3 | 212 | ||
NM_016237.4 | NP_057321.2 | 3 | 434 | |||
XM_005253900.3 | XP_005253957.1 | 3 | 436 | |||
XM_017019423.1 | XP_016874912.1 | 3 | 152 | |||
XM_017019424.1 | XP_016874913.1 | 3 | 152 | |||
GenBank mRNA | AF191339.1 | - | 3 | 312 | ||
AI879332.1 | - | 3 | 436 | |||
AK025614.1 | - | 3 | 212 | |||
AK315451.1 | - | 3 | 380 | |||
BC001081.2 | - | 3 | 367 | 2823401 | ||
BC001950.1 | - | 3 | 367 | 2823401 | ||
BC006301.2 | - | 3 | 371 | 4100632 | ||
BC034243.1 | - | 2 | 74 |