Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137559.1 | NP_001131031.1 | 12 | 1342 | ||
NM_016237.4 | NP_057321.2 | 12 | 1603 | |||
XM_005253900.3 | XP_005253957.1 | 12 | 1566 | |||
XM_017019423.1 | XP_016874912.1 | 11 | 1239 | |||
XM_017019424.1 | XP_016874913.1 | 11 | 1200 | |||
GenBank mRNA | AF191339.1 | - | 12 | 1481 | ||
AK025614.1 | - | 12 | 1342 | |||
AK054779.1 | - | 2 | 1179 | |||
AK294139.1 | - | 5 | 599 | |||
AK301197.1 | - | 4 | 833 | |||
AK315451.1 | - | 12 | 1548 | |||
BC001081.2 | - | 12 | 1536 | 2823401 | ||
BC001950.1 | - | 12 | 1536 | 2823401 | ||
BC003095.1 | - | 7 | 825 | |||
BC006301.2 | - | 12 | 1540 | 4100632 | ||
BC034243.1 | - | 11 | 1243 | |||
BC045686.1 | - | 5 | 1691 | |||
BX537687.1 | - | 7 | 1112 |