Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016255.2 | NP_057339.1 | 2, 3 | 965 | ||
GenBank mRNA | AF097027.1 | - | 2, 3 | 897 | ||
AK296393.1 | - | 3, 4 | 279 | |||
AK307681.1 | - | 2, 3 | 457 | |||
AK312814.1 | - | 2, 3 | 900 | |||
BC047881.1 | - | 2, 3 | 889 | 5742372 |