Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016255.2 | NP_057339.1 | 2 | 925 | ||
GenBank mRNA | AF097027.1 | - | 2 | 857 | ||
AK296393.1 | - | 3 | 239 | |||
AK307681.1 | - | 2 | 417 | |||
AK312814.1 | - | 2 | 860 | |||
BC047881.1 | - | 2 | 849 | 5742372 |