Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016267.3 | NP_057351.1 | 2, 3 | 392 | ||
GenBank mRNA | AF137387.1 | - | 2, 3 | 288 | ||
AK309912.1 | - | 2, 3 | 329 | |||
BC000045.1 | - | 2, 3 | 392 | 3504527 | ||
BC003362.1 | - | 2, 3 | 290 | 2900293 |