Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191324.1 | NP_001178253.2 | 6, 7 | 838 | ||
NM_016271.4 | NP_057355.2 | 7, 8 | 1125 | |||
NM_198128.2 | NP_937761.1 | 4, 5 | 556 | |||
XM_005258285.1 | XP_005258342.1 | 6, 7 | 902 | |||
XM_005258286.1 | XP_005258343.1 | 5, 6 | 843 | |||
GenBank mRNA | AF162680.3 | - | 7, 8 | 888 | ||
AK023579.1 | - | 4, 5 | 474 | |||
AK315761.1 | - | 7, 8 | 1015 | |||
AL133557.1 | - | 5, 6 | 481 | |||
BC018107.1 | - | 7, 8 | 766 | 3914953 | ||
BT006878.1 | - | 6, 7 | 667 | |||
BU662580.1 | - | 4, 5 | 487 | |||
CR457150.1 | - | 6, 7 | 667 |
Gene Symbol: RNF138P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001575.1 | 1 | 4316 | |||
GenBank mRNA | BC040295.1 | - | 1 | 4314 |