Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037872.2 | NP_001032961.1 | 5, 6 | 758 | ||
NM_001321454.1 | NP_001308383.1 | 5, 6 | 758 | |||
NM_001321455.1 | NP_001308384.1 | 6, 7 | 915 | |||
NM_001321458.1 | NP_001308387.1 | 5, 6 | 758 | |||
NM_001321459.1 | NP_001308388.1 | 5, 6 | 758 | |||
NM_001321460.1 | NP_001308389.1 | 5, 6 | 758 | |||
NM_016316.3 | NP_057400.1 | 5, 6 | 758 | |||
NR_135649.1 | 5, 6 | 758 | ||||
NR_135650.1 | 6, 7 | 915 | ||||
NR_135651.1 | 5, 6 | 758 | ||||
NR_135652.1 | 5, 6 | 758 | ||||
NR_135653.1 | 5, 6 | 758 | ||||
XM_017004309.1 | XP_016859798.1 | 5, 6 | 1171 | |||
XM_017004310.1 | XP_016859799.1 | 5, 6 | 537 | |||
XM_017004311.1 | XP_016859800.1 | 5, 6 | 537 | |||
XM_017004312.1 | XP_016859801.1 | 5, 6 | 753 | |||
XM_017004313.1 | XP_016859802.1 | 5, 6 | 2487 | |||
GenBank mRNA | AB047646.1 | - | 5, 6 | 683 | ||
AF151538.1 | - | 5, 6 | 711 | |||
AF206019.1 | - | 5, 6 | 720 | |||
AF357886.1 | - | 5, 6 | 676 | |||
AK002087.1 | - | 3, 4 | 620 | |||
AK307719.1 | - | 5, 6 | 669 | |||
BC037734.1 | - | 6, 7 | 896 | 4824689 | ||
BC130411.1 | - | 4, 5 | 529 | 40146442 | ||
DA852908.1 | - | 2, 3 | 256 | |||
DR000409.1 | - | 648 |