Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037872.2 | NP_001032961.1 | 18 | 3238 | ||
NM_001321454.1 | NP_001308383.1 | 19 | 3349 | |||
NM_001321455.1 | NP_001308384.1 | 19 | 3395 | |||
NM_001321458.1 | NP_001308387.1 | 19 | 3365 | |||
NM_001321459.1 | NP_001308388.1 | 19 | 3441 | |||
NM_001321460.1 | NP_001308389.1 | 19 | 3438 | |||
NM_016316.3 | NP_057400.1 | 18 | 3241 | |||
NR_135649.1 | 19 | 3438 | ||||
NR_135650.1 | 19 | 3395 | ||||
NR_135651.1 | 17 | 3072 | ||||
NR_135652.1 | 18 | 3241 | ||||
NR_135653.1 | 17 | 3075 | ||||
XM_011511341.2 | XP_011509643.1 | 12 | 1858 | |||
XM_017004309.1 | XP_016859798.1 | 19 | 3762 | |||
XM_017004310.1 | XP_016859799.1 | 19 | 3128 | |||
XM_017004311.1 | XP_016859800.1 | 19 | 3128 | |||
XM_017004312.1 | XP_016859801.1 | 19 | 3341 | |||
XM_017004313.1 | XP_016859802.1 | 18 | 4967 | |||
XM_017004314.1 | XP_016859803.1 | 15 | 2618 | |||
XM_017004315.1 | XP_016859804.1 | 15 | 2602 | |||
GenBank mRNA | AB047646.1 | - | 18 | 3163 | ||
AF151538.1 | - | 18 | 3194 | |||
AF206019.1 | - | 18 | 3203 | |||
AF357886.1 | - | 18 | 3159 | |||
AJ131720.1 | - | 10 | 1560 | |||
AK025176.1 | - | 10 | 1520 | |||
AK307719.1 | - | 18 | 3152 | |||
BC037734.1 | - | 19 | 3376 | 4824689 | ||
BC130411.1 | - | 17 | 3009 | 40146442 |