Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2J1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016021.2 | NP_057105.2 | 7 | 956 | ||
XM_011535888.2 | XP_011534190.1 | 7 | 973 | |||
GenBank mRNA | AF151039.1 | - | 4 | 350 | ||
AF151834.1 | - | 7 | 763 | |||
AF161502.1 | - | 7 | 651 | |||
AJ245898.1 | - | 7 | 687 | |||
AJ420578.1 | - | 6 | 605 | |||
AK223464.1 | - | 8 | 695 | |||
AK290574.1 | - | 7 | 798 | |||
AK300629.1 | - | 8 | 701 | |||
BC013973.1 | - | 7 | 918 | 4137664 | ||
U93243.1 | - | 7 | 652 |