Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2J1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016021.2 | NP_057105.2 | 8 | 1058 | ||
XM_011535887.1 | XP_011534189.1 | 7 | 925 | |||
XM_011535888.2 | XP_011534190.1 | 8 | 1105 | |||
GenBank mRNA | AF151039.1 | - | 5 | 452 | ||
AF151834.1 | - | 8 | 865 | |||
AF161502.1 | - | 8 | 753 | |||
AJ245898.1 | - | 8 | 789 | |||
AJ420578.1 | - | 7 | 707 | |||
AK223464.1 | - | 9 | 797 | |||
AK290574.1 | - | 8 | 900 | |||
AK300629.1 | - | 9 | 803 | |||
BC013973.1 | - | 8 | 1020 | 4137664 | ||
U93243.1 | - | 8 | 754 |