Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CABP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318496.1 | NP_001305425.1 | 5 | 550 | ||
NM_016366.2 | NP_057450.2 | 5 | 546 | |||
GenBank mRNA | AB593136.1 | - | 5 | 736 | ||
AF169154.1 | - | 5 | 514 |