Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257999.1 | NP_001244928.1 | 10 | 1543 | ||
NM_016376.3 | NP_057460.3 | 10 | 1338 | |||
NR_047571.1 | 9 | 1219 | ||||
XM_005256679.3 | XP_005256736.1 | 10 | 1385 | |||
XM_011523926.1 | XP_011522228.1 | 10 | 1441 | |||
XM_011523928.2 | XP_011522230.1 | 10 | 1423 | |||
XM_011523933.1 | XP_011522235.1 | 6 | 827 | |||
XM_017024733.1 | XP_016880222.1 | 10 | 1638 | |||
GenBank mRNA | AB033081.2 | - | 10 | 1543 | ||
AB037360.1 | - | 10 | 1300 | |||
AK025483.1 | - | 10 | 1315 | |||
AK057047.1 | - | 9 | 1196 | |||
AK225170.1 | - | 10 | 1315 | |||
AK292930.1 | - | 10 | 1315 | |||
BC052308.1 | - | 10 | 1311 | |||
CR933717.1 | - | 4 | 865 |