Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257999.1 | NP_001244928.1 | 4 | 668 | ||
NM_016376.3 | NP_057460.3 | 4 | 463 | |||
NR_047571.1 | 3 | 344 | ||||
XM_005256679.3 | XP_005256736.1 | 4 | 510 | |||
XM_011523926.1 | XP_011522228.1 | 4 | 566 | |||
XM_011523928.2 | XP_011522230.1 | 4 | 548 | |||
XM_017024733.1 | XP_016880222.1 | 4 | 763 | |||
GenBank mRNA | AB033081.2 | - | 4 | 668 | ||
AB037360.1 | - | 4 | 431 | |||
AK025483.1 | - | 4 | 440 | |||
AK057047.1 | - | 3 | 321 | |||
AK225170.1 | - | 4 | 440 | |||
AK292930.1 | - | 4 | 440 | |||
AK302712.1 | - | 4 | 449 | |||
AL706226.1 | - | 4 | 463 | |||
BC052308.1 | - | 4 | 436 | |||
DA856905.1 | - | 4 | 463 |