Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPOUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016390.3 | NP_057474.2 | 9 | 780 | ||
XM_017014804.1 | XP_016870293.1 | 10 | 1047 | |||
GenBank mRNA | AF161459.1 | - | 9 | 751 | ||
AL110193.1 | - | 9 | 780 | |||
BC010579.1 | - | 9 | 742 | 4213299 | ||
BC021273.1 | - | 9 | 753 | 4843384 | ||
BC033677.1 | - | 9 | 748 | 5176852 | ||
BC039590.1 | - | 9 | 765 | 5757547 | ||
BC046133.1 | - | 9 | 745 | 5725479 | ||
BC063644.1 | - | 9 | 748 | 4539701 |