Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198976.2 | NP_945327.2 | 8, 9 | 1054 | ||
GenBank mRNA | AF161479.1 | - | 8, 9 | 986 | ||
AJ238374.1 | - | 3, 4 | 451 | |||
AJ238375.1 | - | 3, 4 | 377 | |||
AJ238379.1 | - | 8, 9 | 988 | |||
AK001316.1 | - | 8, 9 | 962 | |||
AK023927.1 | - | 8, 9 | 979 | |||
AK293410.1 | - | 8, 9 | 1006 | |||
BC014952.1 | - | 8, 9 | 990 | 4860894 | ||
BX647417.1 | - | 8, 9 | 989 |