Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIKESHI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322404.1 | NP_001309333.1 | 3 | 573 | ||
NM_001322407.1 | NP_001309336.1 | 4 | 747 | |||
NM_001322409.1 | NP_001309338.1 | 4 | 677 | |||
NM_016401.3 | NP_057485.2 | 3 | 589 | |||
NR_024597.1 | 2 | 351 | ||||
NR_024598.1 | 2 | 351 | ||||
NR_136324.1 | 4 | 697 | ||||
XM_017017914.1 | XP_016873403.1 | 3 | 579 | |||
XM_017017915.1 | XP_016873404.1 | 3 | 366 | |||
GenBank mRNA | AA148558.1 | - | 2 | 316 | ||
AF151082.1 | - | 2 | 232 | |||
AF161487.1 | - | 3 | 570 | |||
AF161527.1 | - | 3 | 573 | |||
AK001447.1 | - | 3 | 560 | |||
AK223198.1 | - | 3 | 560 | |||
AK298763.1 | - | 4 | 713 | |||
BC001677.2 | - | 3 | 462 | 2820772 | ||
BC006476.2 | - | 3 | 462 | 2820772 | ||
BC015991.2 | - | 3 | 554 | 2820691 | ||
BC018080.1 | - | 3 | 523 | 4798958 | ||
BC021621.1 | - | 3 | 554 | 2820691 | ||
BP423222.1 | - | 2 | 268 | |||
BX332668.2 | - | 3 | 426 |