Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTSE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016426.6 | NP_057510.4 | 4 | 427 | ||
XM_005261627.3 | XP_005261684.1 | 4 | 1287 | |||
XM_011530211.2 | XP_011528513.1 | 3 | 1229 | |||
XM_011530213.2 | XP_011528515.1 | 4 | 1289 | |||
XM_017028815.1 | XP_016884304.1 | 3 | 677 | |||
XM_017028816.1 | XP_016884305.1 | 4 | 1289 | |||
GenBank mRNA | AF223408.1 | - | 3 | 228 | ||
AK025563.1 | - | 4 | 254 | |||
AK222806.1 | - | 4 | 254 | |||
AK303087.1 | - | 3 | 198 | |||
AK315550.1 | - | 4 | 253 | |||
BC006325.2 | - | 3 | 205 | 4138532 | ||
BC075828.1 | - | 4 | 328 | 5092448 | ||
BG718574.1 | - | 4 | 281 | |||
CT841519.1 | - | 3 | 171 | |||
DA697736.1 | - | 4 | 371 | |||
DB091686.1 | - | 4 | 427 |