Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCKIPSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016453.3 | NP_057537.1 | 9 | 1628 | ||
NM_184231.2 | NP_909119.1 | 9 | 1607 | |||
XM_017006594.1 | XP_016862083.1 | 9 | 1622 | |||
XM_017006595.1 | XP_016862084.1 | 9 | 1615 | |||
XM_017006596.1 | XP_016862085.1 | 9 | 1642 | |||
XM_017006597.1 | XP_016862086.1 | 9 | 1386 | |||
XM_017006598.1 | XP_016862087.1 | 9 | 1612 | |||
XM_017006599.1 | XP_016862088.1 | 9 | 1648 | |||
XM_017006600.1 | XP_016862089.1 | 9 | 1611 | |||
GenBank mRNA | AF178432.1 | - | 9 | 1596 | ||
AF303581.1 | - | 9 | 1567 | |||
AJ242655.2 | - | 9 | 1596 | |||
AK294151.1 | - | 2 | 162 | |||
AY453794.1 | - | 9 | 1575 | |||
BC016052.1 | - | 9 | 1491 | 4862348 | ||
BC026280.1 | - | 9 | 1558 | 4730420 |