Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016464.4 | NP_057548.1 | 5 | 958 | ||
NR_028473.1 | 6 | 841 | ||||
XM_006718585.3 | XP_006718648.1 | 4 | 1196 | |||
XM_006718586.2 | XP_006718649.1 | 5 | 595 | |||
XM_006718588.3 | XP_006718651.1 | 5 | 453 | |||
GenBank mRNA | AF151030.1 | - | 5 | 575 | ||
AF151032.1 | - | 5 | 572 | |||
AK027724.1 | - | 5 | 1107 | |||
AL832879.1 | - | 3 | 3111 | |||
BC005201.1 | - | 5 | 573 | 3684390 | ||
CB956757.1 | - | 6 | 513 |