Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSKIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271904.1 | NP_001258833.1 | 3 | 696 | ||
NM_001271905.1 | NP_001258834.1 | 3 | 249 | |||
NM_001271906.1 | NP_001258835.1 | 2 | 181 | |||
NM_016472.4 | NP_057556.2 | 3 | 282 | |||
XM_017021365.1 | XP_016876854.1 | 3 | 390 | |||
GenBank mRNA | AF151044.1 | - | 3 | 200 | ||
AK000796.1 | - | 3 | 208 | |||
AK094654.1 | - | 3 | 698 | |||
AK314423.1 | - | 3 | 208 | |||
BC004818.2 | - | 2 | 168 | 3449501 | ||
DA488110.1 | - | 3 | 219 | |||
HY019704.1 | - | 3 | 281 | |||
HY051680.1 | - | 3 | 283 | |||
HY108584.1 | - | 2 | 157 |