Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JKAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098625.2 | NP_001092095.1 | 4, 5 | 590 | ||
NM_001284201.1 | NP_001271130.1 | 4, 5 | 1131 | |||
NM_001284202.1 | NP_001271131.1 | 5, 6 | 717 | |||
NM_001284203.1 | NP_001271132.1 | 4, 5 | 640 | |||
NM_001284204.1 | NP_001271133.1 | 4, 5 | 608 | |||
NM_016475.4 | NP_057559.2 | 4, 5 | 608 | |||
XM_006720171.3 | XP_006720234.1 | 4, 5 | 584 | |||
XM_017021366.1 | XP_016876855.1 | 4, 5 | 517 | |||
GenBank mRNA | AF151047.1 | - | 4, 5 | 530 | ||
AF161445.1 | - | 4, 5 | 529 | |||
AF212245.1 | - | 4, 5 | 530 | |||
AF529304.1 | - | 2, 3 | 845 | |||
AK021683.1 | - | 4, 5 | 489 | |||
AK027591.1 | - | 4, 5 | 534 | |||
AK056145.1 | - | 4, 5 | 1007 | |||
AK298209.1 | - | 4, 5 | 515 | |||
AL136696.1 | - | 4, 5 | 500 | |||
BC005198.1 | - | 4, 5 | 516 | 3838401 | ||
BC010359.1 | - | 4, 5 | 547 | 4109351 | ||
BU928810.1 | - | 4, 5 | 538 | |||
BX161501.1 | - | 5, 6 | 600 | |||
CR457163.1 | - | 3, 4 | 443 | |||
CR533510.1 | - | 3, 4 | 461 |
Set Membership: |
G-protein coupled receptor |