Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3HC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282190.1 | NP_001269119.1 | 11 | 1596 | ||
NM_001282191.1 | NP_001269120.1 | 9 | 1337 | |||
NM_016478.4 | NP_057562.3 | 10 | 1550 | |||
XM_005250403.2 | XP_005250460.1 | 8 | 1210 | |||
XM_011516288.2 | XP_011514590.1 | 9 | 1376 | |||
XM_011516289.2 | XP_011514591.1 | 9 | 1761 | |||
XM_011516290.2 | XP_011514592.1 | 8 | 3100 | |||
XM_017012287.1 | XP_016867776.1 | 10 | 1416 | |||
XM_017012288.1 | XP_016867777.1 | 7 | 2052 | |||
GenBank mRNA | AF151050.1 | - | 9 | 1258 | ||
AJ537494.1 | - | 10 | 1465 | |||
AK001317.1 | - | 11 | 1519 | |||
AK022373.1 | - | 10 | 1473 | |||
BC000190.1 | - | 10 | 1458 | |||
BC011551.1 | - | 10 | 1466 | 4561164 | ||
BC028917.1 | - | 5 | 742 |