Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3HC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282190.1 | NP_001269119.1 | 4 | 435 | ||
NM_001282191.1 | NP_001269120.1 | 3 | 389 | |||
NM_016478.4 | NP_057562.3 | 3 | 389 | |||
XM_017012287.1 | XP_016867776.1 | 3 | 384 | |||
GenBank mRNA | AF151050.1 | - | 3 | 310 | ||
AJ537494.1 | - | 3 | 304 | |||
AK001317.1 | - | 4 | 358 | |||
AK022373.1 | - | 3 | 312 | |||
BC000190.1 | - | 3 | 297 | |||
BC011551.1 | - | 3 | 305 | 4561164 | ||
BI830062.1 | - | 3 | 391 | |||
BM013594.1 | - | 3 | 307 | |||
DA066102.1 | - | 3 | 346 |