Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286371.1 | NP_001273300.1 | 2 | 271 | ||
NM_016485.4 | NP_057569.2 | 2 | 271 | |||
GenBank mRNA | AF060225.1 | - | 2 | 161 | ||
AF141341.1 | - | 2 | 148 | |||
AF151062.1 | - | 2 | 174 | |||
AF271994.1 | - | 2 | 160 | |||
AJ420507.1 | - | 2 | 166 | |||
AK000051.1 | - | 2 | 160 | |||
AK130738.1 | - | 2 | 163 | |||
AL136684.1 | - | 2 | 164 | |||
BC005937.1 | - | 2 | 156 | 4081455 | ||
BC006989.1 | - | 2 | 170 | 3961637 | ||
BC022536.1 | - | 2 | 170 | 4817057 | ||
BM806661.1 | - | 2 | 71 | |||
CR457207.1 | - | 2 | 145 | |||
DA299959.1 | - | 2 | 271 |