Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293212.1 | NP_001280141.1 | 4 | 841 | ||
NM_001293214.1 | NP_001280143.1 | 3 | 625 | |||
NM_001293215.1 | NP_001280144.1 | 4 | 744 | |||
NM_001293216.1 | NP_001280145.1 | 4 | 643 | |||
NM_178014.3 | NP_821133.1 | 4 | 757 | |||
GenBank mRNA | AB062393.1 | - | 4 | 583 | ||
AF070561.1 | - | 4 | 578 | |||
AF070593.1 | - | 4 | 578 | |||
AF070600.1 | - | 4 | 595 | |||
AF141349.1 | - | 4 | 595 | |||
AK098772.1 | - | 4 | 602 | |||
AK298889.1 | - | 4 | 647 | |||
AK302314.1 | - | 4 | 710 | |||
AK303227.1 | - | 3 | 344 | |||
AK309684.1 | - | 2 | 199 | |||
AK316265.1 | - | 4 | 514 | |||
AK316316.1 | - | 4 | 639 | |||
BC001896.1 | - | 1 | 139 | |||
BC001938.1 | - | 4 | 582 | 2820585 | ||
BC002347.2 | - | 4 | 587 | 2964461 | ||
BC005838.2 | - | 4 | 582 | 2820585 | ||
BC007605.1 | - | 4 | 587 | 3350604 | ||
BC013374.2 | - | 4 | 578 | 3946253 | ||
BC019924.1 | - | 4 | 575 | 4998571 | ||
BC020946.1 | - | 4 | 616 | 4795692 | ||
BC021909.1 | - | 4 | 577 | 3509417 | ||
BC070326.1 | - | 4 | 593 | 6698679 | ||
BC103746.1 | - | 4 | 600 | 6503977 | ||
BU557299.1 | - | 4 | 757 | |||
DC389555.1 | - | 3 | 469 | |||
DC405540.1 | - | 4 | 554 | |||
DQ457058.1 | - | 1 | 13 | |||
V00599.1 | - | 3 | 430 |