Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016538.2 | NP_057622.1 | 9 | 986 | ||
XM_006722284.2 | XP_006722347.1 | 7 | 720 | |||
XM_011523580.1 | XP_011521882.1 | 8 | 852 | |||
XM_011523581.2 | XP_011521883.1 | 8 | 2589 | |||
GenBank mRNA | AF233395.1 | - | 9 | 968 | ||
AK094326.1 | - | 8 | 1051 | |||
AK131437.1 | - | 5 | 2169 | |||
AK131484.1 | - | 4 | 1541 | |||
AK290265.1 | - | 9 | 954 | |||
AL137626.1 | - | 6 | 554 | |||
BC017305.2 | - | 9 | 950 | 5087554 | ||
BC101791.1 | - | 9 | 967 | 8069297 | ||
BC101793.1 | - | 9 | 966 | 8069299 |