Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016577.3 | NP_057661.3 | 3 | 523 | ||
XM_011512893.2 | XP_011511195.1 | 3 | 198 | |||
GenBank mRNA | AF091031.1 | - | 2 | 70 | ||
AF166492.1 | - | 3 | 479 | |||
AF498940.1 | - | 3 | 142 | |||
AK295451.1 | - | 4 | 260 | |||
AK312378.1 | - | 3 | 443 | |||
BC002510.2 | - | 3 | 492 | 3050592 | ||
BC078662.1 | - | 3 | 271 | 6450986 | ||
BT007263.1 | - | 3 | 142 |