Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144891.1 | NP_001138363.1 | 6 | 834 | ||
NM_001308110.1 | NP_001295039.1 | 6 | 834 | |||
NM_016586.2 | NP_057670.2 | 6 | 834 | |||
XM_005267754.3 | XP_005267811.1 | 5 | 763 | |||
XM_005267756.4 | XP_005267813.1 | 6 | 829 | |||
XM_017021367.1 | XP_016876856.1 | 5 | 763 | |||
GenBank mRNA | AB038523.1 | - | 6 | 803 | ||
AF208857.1 | - | 6 | 783 | |||
AK293473.1 | - | 6 | 715 | |||
AK315290.1 | - | 6 | 835 | |||
BC005197.1 | - | 6 | 793 | 3681897 | ||
BC016821.1 | - | 6 | 789 | 4097046 | ||
BX248287.1 | - | 6 | 754 |