Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016614.2 | NP_057698.2 | 7 | 950 | ||
GenBank mRNA | AF201687.1 | - | 7 | 950 | ||
AF223469.1 | - | 7 | 941 | |||
AJ269473.1 | - | 7 | 946 | |||
AJ420495.1 | - | 6 | 1151 | |||
AK002168.1 | - | 7 | 925 | |||
AK296623.1 | - | 6 | 626 | |||
AK298699.1 | - | 7 | 1155 | |||
BC017553.2 | - | 7 | 962 | 3920790 | ||
BC110375.1 | - | 7 | 931 | 5737150 |