Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016614.2 | NP_057698.2 | 4 | 519 | ||
GenBank mRNA | AF201687.1 | - | 4 | 519 | ||
AF223469.1 | - | 4 | 510 | |||
AJ269473.1 | - | 4 | 515 | |||
AJ420495.1 | - | 3 | 720 | |||
AK002168.1 | - | 4 | 494 | |||
AK296623.1 | - | 3 | 195 | |||
AK298699.1 | - | 4 | 724 | |||
AK308836.1 | - | 4 | 522 | |||
BC017553.2 | - | 4 | 531 | 3920790 | ||
BC110375.1 | - | 4 | 500 | 5737150 |