Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286703.1 | NP_001273632.1 | 4 | 291 | ||
NM_001286704.1 | NP_001273633.1 | 4 | 469 | |||
NM_001286705.1 | NP_001273634.1 | 4 | 297 | |||
NM_001286706.1 | NP_001273635.1 | 4 | 297 | |||
NM_016617.3 | NP_057701.1 | 4 | 297 | |||
NR_104584.1 | 5 | 358 | ||||
GenBank mRNA | AB154404.1 | - | 3 | 148 | ||
AF208844.1 | - | 4 | 187 | |||
AK095490.1 | - | 4 | 217 | |||
AK307111.1 | - | 4 | 217 | |||
AK309914.1 | - | 4 | 183 | |||
AL515797.3 | - | 3 | 149 | |||
BC005193.1 | - | 4 | 214 | 3829206 | ||
BG533062.1 | - | 4 | 390 | |||
BG540094.1 | - | 4 | 185 | |||
BI458294.1 | - | 4 | 268 | |||
BI761948.1 | - | 4 | 174 | |||
BP277186.1 | - | 4 | 297 | |||
BU589105.1 | - | 5 | 274 | |||
CR457189.1 | - | 3 | 148 | |||
DA483023.1 | - | 4 | 217 | |||
Z70222.1 | - | 4 | 188 |