Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LARP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267039.1 | NP_001253968.1 | 4 | 653 | ||
NM_015454.2 | NP_056269.1 | 2 | 317 | |||
NM_016648.3 | NP_057732.2 | 2 | 445 | |||
GenBank mRNA | AK000274.1 | - | 2 | 255 | ||
BC006981.1 | - | 2 | 228 | |||
BC066945.1 | - | 4 | 491 | 4837780 | ||
CN346208.1 | - | 2 | 324 | |||
DB270203.1 | - | 2 | 445 | |||
EU667388.1 | - | 2 | 188 |