Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LARP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267039.1 | NP_001253968.1 | 11 | 1744 | ||
NM_015454.2 | NP_056269.1 | 9 | 1408 | |||
NM_016648.3 | NP_057732.2 | 9 | 1536 | |||
NR_049768.1 | 9 | 1433 | ||||
GenBank mRNA | AF068284.1 | - | 8 | 685 | ||
AK000274.1 | - | 9 | 1346 | |||
AL049996.1 | - | 3 | 383 | |||
BC066945.1 | - | 11 | 1582 | 4837780 | ||
BC107709.1 | - | 3 | 377 | |||
BX647100.1 | - | 9 | 1270 | |||
EU667388.1 | - | 9 | 1279 |