Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015885.3 | NP_056969.2 | 10 | 4124 | ||
XM_005274048.1 | XP_005274105.1 | 10 | 4517 | |||
XM_005274049.1 | XP_005274106.1 | 10 | 4517 | |||
XM_017017918.1 | XP_016873407.1 | 10 | 4124 | |||
GenBank mRNA | AB020631.1 | - | 10 | 4048 | ||
BC065384.1 | - | 10 | 4016 | 6043635 | ||
BC146778.1 | - | 10 | 4048 | 9007228 |