Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014433.2 | NP_001014433.1 | 4, 5 | 523 | ||
NM_001014837.1 | NP_001014837.1 | 4, 5 | 772 | |||
NM_001014838.1 | NP_001014838.1 | 4, 5 | 741 | |||
NM_001014840.1 | NP_001014840.1 | 4, 5 | 651 | |||
NM_015921.2 | NP_057005.1 | 3, 4 | 914 | |||
GenBank mRNA | AF106943.1 | - | 3, 4 | 551 | ||
AF230924.1 | - | 3, 4 | 337 | |||
BC005890.2 | - | 4, 5 | 348 | 3505574 | ||
BC107751.1 | - | 3, 4 | 551 | 6714433 | ||
CN363642.1 | - | 4, 5 | 553 |