Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007239.1 | NP_001007240.1 | 2 | 570 | ||
NM_014955.2 | NP_055770.1 | 2 | 245 | |||
NM_015935.4 | NP_057019.3 | 2 | 570 | |||
XM_017001455.1 | XP_016856944.1 | 1 | 64 | |||
XM_017001456.1 | XP_016856945.1 | 1 | 64 | |||
XM_017001457.1 | XP_016856946.1 | 1 | 64 | |||
XM_017001458.1 | XP_016856947.1 | 2 | 687 | |||
XM_017001459.1 | XP_016856948.1 | 2 | 237 | |||
GenBank mRNA | AB020666.2 | - | 2 | 325 | ||
AB242174.1 | - | 2 | 495 | |||
AB242175.1 | - | 2 | 483 | |||
AF132936.1 | - | 2 | 495 | |||
AK027552.1 | - | 2 | 489 | |||
AK027621.1 | - | 2 | 184 | |||
AK027886.1 | - | 1 | 41 | |||
AK223493.1 | - | 2 | 531 | |||
AK291740.1 | - | 2 | 517 | |||
AK304471.1 | - | 3 | 351 | |||
AL049669.1 | - | 2 | 495 | |||
BC029083.1 | - | 2 | 324 | 4509086 |