Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007239.1 | NP_001007240.1 | 7 | 1590 | ||
NM_014955.2 | NP_055770.1 | 7 | 1733 | |||
NM_015935.4 | NP_057019.3 | 7 | 2058 | |||
XM_017001455.1 | XP_016856944.1 | 6 | 1552 | |||
XM_017001456.1 | XP_016856945.1 | 6 | 1552 | |||
XM_017001457.1 | XP_016856946.1 | 5 | 1333 | |||
XM_017001458.1 | XP_016856947.1 | 6 | 2010 | |||
XM_017001459.1 | XP_016856948.1 | 6 | 1560 | |||
GenBank mRNA | AB020666.2 | - | 7 | 1813 | ||
AB242174.1 | - | 7 | 1983 | |||
AB242175.1 | - | 7 | 1971 | |||
AF132936.1 | - | 7 | 1983 | |||
AK001172.1 | - | 5 | 745 | |||
AK027552.1 | - | 7 | 1977 | |||
AK027621.1 | - | 7 | 1672 | |||
AK027886.1 | - | 6 | 1529 | |||
AK074552.1 | - | 3 | 811 | |||
AK129561.1 | - | 6 | 1406 | |||
AK223493.1 | - | 7 | 2019 | |||
AK291740.1 | - | 7 | 2005 | |||
AK304471.1 | - | 8 | 1839 | |||
AL049669.1 | - | 7 | 1515 | |||
BC006814.1 | - | 4 | 608 | |||
BC029083.1 | - | 7 | 1812 | 4509086 |