Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293212.1 | NP_001280141.1 | 4 | 856 | ||
NM_001293214.1 | NP_001280143.1 | 3 | 640 | |||
NM_001293215.1 | NP_001280144.1 | 4 | 759 | |||
NM_001293216.1 | NP_001280145.1 | 4 | 658 | |||
NM_178014.3 | NP_821133.1 | 4 | 772 | |||
GenBank mRNA | AB062393.1 | - | 4 | 598 | ||
AF070561.1 | - | 4 | 593 | |||
AF070593.1 | - | 4 | 593 | |||
AF070600.1 | - | 4 | 610 | |||
AF141349.1 | - | 4 | 610 | |||
AK098772.1 | - | 4 | 617 | |||
AK298889.1 | - | 4 | 662 | |||
AK302314.1 | - | 4 | 725 | |||
AK303227.1 | - | 3 | 359 | |||
AK309684.1 | - | 2 | 214 | |||
AK316265.1 | - | 4 | 529 | |||
AK316316.1 | - | 4 | 654 | |||
BC001896.1 | - | 1 | 154 | |||
BC001938.1 | - | 4 | 597 | 2820585 | ||
BC002347.2 | - | 4 | 602 | 2964461 | ||
BC005838.2 | - | 4 | 597 | 2820585 | ||
BC007605.1 | - | 4 | 602 | 3350604 | ||
BC013374.2 | - | 4 | 593 | 3946253 | ||
BC019924.1 | - | 4 | 590 | 4998571 | ||
BC020946.1 | - | 4 | 631 | 4795692 | ||
BC021909.1 | - | 4 | 592 | 3509417 | ||
BC070326.1 | - | 4 | 608 | 6698679 | ||
BC103746.1 | - | 4 | 615 | 6503977 | ||
BU557299.1 | - | 4 | 772 | |||
DC389555.1 | - | 3 | 484 | |||
DQ457058.1 | - | 1 | 28 | |||
V00599.1 | - | 3 | 445 |