Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077394.1 | NP_001070862.1 | 3 | 332 | ||
NM_001077395.1 | NP_001070863.1 | 3 | 314 | |||
NM_015958.2 | NP_057042.2 | 3 | 314 | |||
XM_005270938.1 | XP_005270995.1 | 3 | 314 | |||
XM_005270939.4 | XP_005270996.1 | 2 | 122 | |||
XM_005270941.3 | XP_005270998.1 | 3 | 249 | |||
XM_006710683.3 | XP_006710746.1 | 2 | 121 | |||
XM_011541569.1 | XP_011539871.1 | 3 | 314 | |||
XM_011541570.2 | XP_011539872.1 | 2 | 114 | |||
XM_017001460.1 | XP_016856949.1 | 3 | 334 | |||
XM_017001461.1 | XP_016856950.1 | 2 | 121 | |||
XM_017001462.1 | XP_016856951.1 | 3 | 271 | |||
GenBank mRNA | AF132964.1 | - | 3 | 283 | ||
AF157319.1 | - | 3 | 282 | |||
AF161492.1 | - | 3 | 323 | |||
AF248965.1 | - | 3 | 283 | |||
AK125485.1 | - | 4 | 683 | |||
AK225469.1 | - | 3 | 292 | |||
AK225707.1 | - | 3 | 292 | |||
AK225951.1 | - | 3 | 292 | |||
AK289351.1 | - | 3 | 303 | |||
AK298181.1 | - | 3 | 292 | |||
BC053857.1 | - | 3 | 280 | 5503038 | ||
BI907367.1 | - | 3 | 287 | |||
CK002046.1 | - | 3 | 335 | |||
DA133565.1 | - | 3 | 314 |