Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERGIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015966.2 | NP_057050.1 | 2 | 210 | ||
NM_198398.1 | NP_938408.1 | 2 | 210 | |||
GenBank mRNA | AF077030.1 | - | 2 | 168 | ||
AF116614.1 | - | 2 | 317 | |||
AF151812.1 | - | 2 | 168 | |||
AF308298.1 | - | 2 | 170 | |||
AK074550.1 | - | 2 | 158 | |||
AK096484.1 | - | 2 | 158 | |||
AK300916.1 | - | 2 | 159 | |||
BC009765.1 | - | 2 | 164 | 4283844 | ||
BC014014.2 | - | 2 | 163 | 3954269 | ||
BU528333.1 | - | 2 | 187 | |||
CA489637.1 | - | 2 | 210 | |||
EF206690.1 | - | 2 | 168 |