Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016024.2 | NP_057108.2 | 6 | 930 | ||
GenBank mRNA | AF151837.1 | - | 5 | 865 | ||
AK292855.1 | - | 6 | 912 | |||
AL050405.1 | - | 6 | 905 | |||
BC033750.1 | - | 6 | 898 | 5210341 | ||
BC040433.1 | - | 6 | 881 |